Gene Transcripts

Gene Symbol
Pou5f2
Entrez Gene ID
75507
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
1700013G10Rik, SPRM-1, Sprm1
Chromosome Location
Chr. 13: 78024902 - 78026296 on Build GRCm38
UniGene ID
Mm.443255
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029315.1 NP_083591.1 1 1152
GenBank mRNA AK005948.1 - 1 1153
AK076753.1 - 1 1143

View Full Product Details