Gene Transcripts
- Gene Symbol
- ELF3
- Entrez Gene ID
- 1999
- Gene Name
- E74 like ETS transcription factor 3
- Gene Aliases
- EPR-1, ERT, ESE-1, ESX
- Chromosome Location
- Chr. 1: 202010519 - 202017188 on Build GRCh38
- UniGene ID
- Hs.603657
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114309.1 |
NP_001107781.1
|
8, 9 |
1276 |
|
NM_004433.4 |
NP_004424.3
|
8, 9 |
1162 |
|
GenBank mRNA |
AF016295.1 |
- |
8, 9 |
1123 |
|
AF017307.1 |
- |
8, 9 |
1211 |
|
AF517841.1 |
- |
8, 9 |
1144 |
|
AK225446.1 |
- |
8, 9 |
1099 |
|
AK310814.1 |
- |
8, 9 |
1154 |
|
AK312273.1 |
- |
8, 9 |
1138 |
|
BC003569.1 |
- |
8, 9 |
1130 |
3537846 |
BX537368.1 |
- |
7, 8 |
4199 |
|
CR457106.1 |
- |
7, 8 |
1009 |
|
U66894.1 |
- |
8, 9 |
1104 |
|
U73843.1 |
- |
8, 9 |
1128 |
|
U73844.1 |
- |
9, 10 |
1059 |
|
U97156.1 |
- |
8, 9 |
1133 |
|
View Full Product Details