Gene Transcripts

Gene Symbol
Prelid2
Entrez Gene ID
77619
Gene Name
PRELI domain containing 2
Gene Aliases
1700003A01Rik, C330008K14Rik
Chromosome Location
Chr. 18: 41875696 - 41951194 on Build GRCm38
UniGene ID
Mm.30649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029942.1 NP_084218.1 5, 6 503
GenBank mRNA AK021193.1 - 5, 6 529
BC119251.1 - 5, 6 503 8734000
BC120711.1 - 5, 6 503 40129634

View Full Product Details