Gene Transcripts
- Gene Symbol
- Neurod2
- Entrez Gene ID
- 54276
- Gene Name
- neuronal differentiation 2
- Gene Aliases
- Chromosome Location
- Chr. 10: 86280865 - 86282681 on Build Rnor_6.0
- UniGene ID
- Rn.10724
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019326.1 |
NP_062199.1
|
1 |
1155 |
|
GenBank mRNA |
D82868.1 |
- |
1 |
1155 |
|
View Full Product Details