Gene Transcripts

Gene Symbol
Slc9a3r1
Entrez Gene ID
59114
Gene Name
solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1
Gene Aliases
Chromosome Location
Chr. 10: 103713045 - 103730145 on Build Rnor_6.0
UniGene ID
Rn.35142
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021594.1 NP_067605.1 3 767
GenBank mRNA AF154336.1 - 3 767
FQ213792.1 - 3 884
FQ228926.1 - 3 865

View Full Product Details