Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004271.3 | NP_004262.1 | 3 | 313 | |
XM_017011505.1 | XP_016866994.1 | 3 | 375 | ||
GenBank mRNA | AB020499.1 | - | 2 | 149 | |
AF057178.2 | - | 3 | 302 | ||
AY931019.1 | - | 3 | 279 | ||
BC038846.1 | - | 3 | 323 | 6015123 | |
CD695164.1 | - | 3 | 377 |