Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021854.1 | NP_068626.1 | 4, 5 | 516 | |
XM_006233843.2 | XP_006233905.1 | 6, 7 | 970 | ||
XM_006233844.2 | XP_006233906.1 | 6, 7 | 941 | ||
XM_006233845.2 | XP_006233907.1 | 6, 7 | 792 | ||
XM_006233846.2 | XP_006233908.1 | 6, 7 | 3057 | ||
XM_006233847.2 | XP_006233909.1 | 6, 7 | 1278 | ||
XM_006233849.2 | XP_006233911.1 | 6, 7 | 970 | ||
GenBank mRNA | AB011821.1 | - | 4, 5 | 516 |