Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022205.3 | NP_071541.2 | 2 | 404 | |
GenBank mRNA | AF452185.1 | - | 1 | 329 | |
BC089804.1 | - | 2 | 384 | 7380432 | |
CO557829.1 | - | 2 | 404 | ||
U54791.1 | - | 2 | 366 | ||
U90610.1 | - | 1 | 329 |