Gene Transcripts

Gene Symbol
Ptprq
Entrez Gene ID
360417
Gene Name
protein tyrosine phosphatase, receptor type, Q
Gene Aliases
rPTP-GMC1
Chromosome Location
Chr. 7: 49763657 - 50034932 on Build Rnor_6.0
UniGene ID
Rn.30011
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022925.1 NP_075214.1 3 472
GenBank mRNA AF063249.1 - 3 472

Gene Symbol
LOC102552564
Entrez Gene ID
102552564
Gene Name
phosphatidylinositol phosphatase PTPRQ-like
Gene Aliases
Chromosome Location
Chr. 7: 51986133 - 51990630 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241319.2 XP_006241381.1 3 476

View Full Product Details