Gene Transcripts
- Gene Symbol
- Axin2
- Entrez Gene ID
- 29134
- Gene Name
- axin 2
- Gene Aliases
- axil
- Chromosome Location
- Chr. 10: 97212483 - 97238824 on Build Rnor_6.0
- UniGene ID
- Rn.162212
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024355.1 |
NP_077331.1
|
9, 10 |
2471 |
|
GenBank mRNA |
AF017757.1 |
- |
9, 10 |
2471 |
|
View Full Product Details