Gene Transcripts

Gene Symbol
GSTO1
Entrez Gene ID
9446
Gene Name
glutathione S-transferase omega 1
Gene Aliases
GSTO 1-1, GSTTLp28, HEL-S-21, P28, SPG-R
Chromosome Location
Chr. 10: 104254194 - 104267464 on Build GRCh38
UniGene ID
Hs.190028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191002.1 NP_001177931.1 2, 3 356
NM_001191003.1 NP_001177932.1 2, 3 409
NM_004832.2 NP_004823.1 2, 3 356
GenBank mRNA AF212303.1 - 2, 3 163
AK313680.1 - 2, 3 174
BC000127.1 - 2, 3 166 3351359
BX537431.1 - 2, 3 284
CF130513.1 - 2, 3 133
CN283947.1 - 2, 3 159
DB501888.1 - 2, 3 440
DT220068.1 - 2, 3 391
EU794619.1 - 2, 3 163
U90313.1 - 2, 3 146

View Full Product Details