Gene Transcripts

Gene Symbol
Sctr
Entrez Gene ID
81779
Gene Name
secretin receptor
Gene Aliases
Chromosome Location
Chr. 13: 36039241 - 36090515 on Build Rnor_6.0
UniGene ID
Rn.32256
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031115.1 NP_112377.1 8 1050
GenBank mRNA BC081781.1 - 8 1008
X59132.1 - 8 1050

View Full Product Details