Gene Transcripts
- Gene Symbol
- EPHB6
- Entrez Gene ID
- 2051
- Gene Name
- EPH receptor B6
- Gene Aliases
- HEP
- Chromosome Location
- Chr. 7: 142854640 - 142871093 on Build GRCh38
- UniGene ID
- Hs.380089
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001280794.2 |
NP_001267723.2
|
9, 10 |
2333 |
|
NM_001280795.2 |
NP_001267724.2
|
6, 7 |
1705 |
|
NM_004445.5 |
NP_004436.4
|
7, 8 |
1774 |
|
XM_006715881.2 |
XP_006715944.1
|
10, 11 |
2769 |
|
XM_011515879.1 |
XP_011514181.1
|
11, 12 |
2808 |
|
XM_011515880.1 |
XP_011514182.1
|
10, 11 |
2786 |
|
XM_011515881.2 |
XP_011514183.1
|
10, 11 |
2770 |
|
GenBank mRNA |
BC056159.1 |
- |
9, 10 |
2202 |
|
BC110606.1 |
- |
6, 7 |
1699 |
40028676 |
BC110607.1 |
- |
7, 8 |
1768 |
40028687 |
D83492.1 |
- |
10, 11 |
2345 |
|
EU054308.1 |
- |
5, 6 |
1711 |
|
View Full Product Details