Gene Transcripts

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
Chromosome Location
Chr. 7: 142854640 - 142871093 on Build GRCh38
UniGene ID
Hs.380089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280794.2 NP_001267723.2 13, 14 2771
NM_001280795.2 NP_001267724.2 10, 11 2143
NM_004445.5 NP_004436.4 11, 12 2212
NR_104001.2 13, 14 2023
XM_006715881.2 XP_006715944.1 14, 15 3207
XM_011515879.1 XP_011514181.1 15, 16 3246
XM_011515880.1 XP_011514182.1 14, 15 3224
XM_011515881.2 XP_011514183.1 14, 15 3208
XM_011515882.2 XP_011514184.1 14, 15 3117
GenBank mRNA AL832533.1 - 1, 2 1074
BC056159.1 - 13, 14 2640
BC110606.1 - 10, 11 2137 40028676
BC110607.1 - 11, 12 2206 40028687
D83492.1 - 14, 15 2783
EU054308.1 - 9, 10 2149

View Full Product Details