Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031632.1 | NP_113820.1 | 2 | 304 | |
GenBank mRNA | AF262319.1 | - | 2 | 304 | |
AF271996.1 | - | 2 | 271 | ||
AF286006.1 | - | 2 | 304 | ||
AF293333.1 | - | 2 | 304 | ||
AF308469.1 | - | 2 | 271 | ||
AF517560.1 | - | 2 | 304 | ||
AY008275.1 | - | 2 | 271 | ||
AY027667.1 | - | 2 | 280 | ||
AY124461.1 | - | 2 | 304 |