Gene Transcripts

Gene Symbol
Ltb4r2
Entrez Gene ID
114098
Gene Name
leukotriene B4 receptor 2
Gene Aliases
Chromosome Location
Chr. 15: 34446478 - 34448183 on Build Rnor_6.0
UniGene ID
Rn.74251
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053640.1 NP_446092.1 1 117
XM_008770658.1 XP_008768880.1 1 137
GenBank mRNA AB052660.1 - 1 117

View Full Product Details