Gene Transcripts

Gene Symbol
Slc9a3r2
Entrez Gene ID
116501
Gene Name
solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2
Gene Aliases
E3karp
Chromosome Location
Chr. 10: 14003330 - 14013844 on Build Rnor_6.0
UniGene ID
Rn.39351
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053811.3 NP_446263.2 2 353
GenBank mRNA AF259898.1 - 2 212
AF294257.1 - 2 440
CK840521.1 - 2 140

View Full Product Details