Gene Transcripts
- Gene Symbol
- Rps23
- Entrez Gene ID
- 124323
- Gene Name
- ribosomal protein S23
- Gene Aliases
- Chromosome Location
- Chr. 2: 19823234 - 19824804 on Build Rnor_6.0
- UniGene ID
- Rn.2139
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_078617.3 |
NP_511172.1
|
3, 4 |
332 |
|
GenBank mRNA |
BC058134.1 |
- |
3, 4 |
336 |
|
FQ221811.1 |
- |
3, 4 |
324 |
|
FQ222488.1 |
- |
3, 4 |
322 |
|
FQ222494.1 |
- |
3, 4 |
326 |
|
FQ222496.1 |
- |
3, 4 |
323 |
|
FQ222641.1 |
- |
3, 4 |
324 |
|
FQ223677.1 |
- |
3, 4 |
324 |
|
FQ223812.1 |
- |
3, 4 |
324 |
|
FQ228659.1 |
- |
3, 4 |
324 |
|
FQ228931.1 |
- |
3, 4 |
324 |
|
View Full Product Details