Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134326.2 | NP_599153.2 | 8 | 1034 | |
GenBank mRNA | BC085359.1 | - | 8 | 1034 | |
FQ209382.1 | - | 4 | 482 | ||
FQ209565.1 | - | 1 | 67 | ||
FQ209580.1 | - | 1 | 19 | ||
FQ209635.1 | - | 2 | 244 | ||
FQ209651.1 | - | 2 | 274 | ||
FQ209693.1 | - | 1 | 69 | ||
FQ209714.1 | - | 1 | 137 | ||
FQ209721.1 | - | 2 | 255 | ||
FQ209825.1 | - | 1 | 60 | ||
FQ209907.1 | - | 4 | 503 | ||
FQ209957.1 | - | 3 | 415 | ||
FQ209968.1 | - | 1 | 62 | ||
FQ210022.1 | - | 2 | 257 | ||
FQ210057.1 | - | 4 | 433 | ||
FQ210111.1 | - | 2 | 286 | ||
FQ210171.1 | - | 2 | 255 | ||
FQ210297.1 | - | 1 | 102 | ||
FQ210310.1 | - | 6 | 706 | ||
FQ210316.1 | - | 2 | 255 | ||
FQ210347.1 | - | 1 | 36 | ||
FQ210361.1 | - | 1 | 175 | ||
FQ210406.1 | - | 2 | 234 | ||
FQ210411.1 | - | 1 | 166 | ||
FQ210546.1 | - | 2 | 282 | ||
FQ210564.1 | - | 1 | 19 | ||
FQ210640.1 | - | 2 | 239 | ||
FQ210724.1 | - | 5 | 558 | ||
FQ210803.1 | - | 4 | 504 | ||
FQ210850.1 | - | 3 | 350 | ||
FQ210851.1 | - | 3 | 381 | ||
FQ210858.1 | - | 5 | 588 | ||
FQ210877.1 | - | 4 | 449 | ||
FQ210995.1 | - | 2 | 244 | ||
FQ211001.1 | - | 1 | 74 | ||
FQ211010.1 | - | 3 | 392 | ||
FQ211036.1 | - | 2 | 287 | ||
FQ211096.1 | - | 2 | 255 | ||
FQ211129.1 | - | 2 | 228 | ||
FQ211196.1 | - | 2 | 215 | ||
FQ211218.1 | - | 5 | 662 | ||
FQ211255.1 | - | 4 | 482 | ||
FQ211259.1 | - | 4 | 491 | ||
FQ211320.1 | - | 2 | 286 | ||
FQ211355.1 | - | 1 | 79 | ||
FQ211378.1 | - | 2 | 255 | ||
FQ211445.1 | - | 1 | 159 | ||
FQ218069.1 | - | 5 | 494 | ||
FQ218081.1 | - | 1 | 63 | ||
FQ218137.1 | - | 2 | 259 | ||
FQ218216.1 | - | 3 | 358 | ||
FQ218412.1 | - | 1 | 66 | ||
FQ218458.1 | - | 4 | 498 | ||
FQ218566.1 | - | 1 | 165 | ||
FQ218682.1 | - | 2 | 223 | ||
FQ218739.1 | - | 3 | 403 | ||
FQ218815.1 | - | 3 | 368 | ||
FQ218867.1 | - | 1 | 184 | ||
FQ218944.1 | - | 1 | 97 | ||
FQ218989.1 | - | 1 | 88 | ||
FQ219017.1 | - | 1 | 39 | ||
FQ219210.1 | - | 4 | 453 | ||
FQ219231.1 | - | 1 | 80 | ||
FQ219260.1 | - | 2 | 224 | ||
FQ219293.1 | - | 4 | 482 | ||
FQ219382.1 | - | 1 | 169 | ||
FQ219442.1 | - | 2 | 252 | ||
FQ219447.1 | - | 4 | 475 | ||
FQ219470.1 | - | 3 | 353 | ||
FQ219477.1 | - | 1 | 95 | ||
FQ219514.1 | - | 2 | 281 | ||
FQ219650.1 | - | 2 | 251 | ||
FQ219670.1 | - | 4 | 532 | ||
FQ219679.1 | - | 2 | 244 | ||
FQ219682.1 | - | 2 | 226 | ||
FQ219734.1 | - | 6 | 686 | ||
FQ219775.1 | - | 1 | 138 | ||
FQ224096.1 | - | 1 | 138 | ||
V01222.1 | - | 8 | 1019 |