Gene Transcripts
- Gene Symbol
- Apoe
- Entrez Gene ID
- 25728
- Gene Name
- apolipoprotein E
- Gene Aliases
- APOEA
- Chromosome Location
- Chr. 1: 80612894 - 80616820 on Build Rnor_6.0
- UniGene ID
- Rn.32351
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270681.1 |
NP_001257610.1
|
3, 4 |
348 |
|
NM_001270682.1 |
NP_001257611.1
|
3, 4 |
327 |
|
NM_001270683.1 |
NP_001257612.1
|
3, 4 |
314 |
|
NM_001270684.1 |
NP_001257613.1
|
3, 4 |
290 |
|
NM_138828.3 |
NP_620183.2
|
3, 4 |
308 |
|
GenBank mRNA |
BC060313.1 |
- |
3, 4 |
281 |
|
BC086581.1 |
- |
3, 4 |
251 |
7307758 |
FQ209421.1 |
- |
3, 4 |
273 |
|
FQ209426.1 |
- |
3, 4 |
279 |
|
FQ209428.1 |
- |
3, 4 |
277 |
|
FQ209457.1 |
- |
3, 4 |
276 |
|
FQ209625.1 |
- |
3, 4 |
277 |
|
FQ209879.1 |
- |
2, 3 |
224 |
|
FQ209926.1 |
- |
3, 4 |
277 |
|
FQ210262.1 |
- |
3, 4 |
277 |
|
FQ210410.1 |
- |
3, 4 |
277 |
|
FQ211250.1 |
- |
3, 4 |
278 |
|
FQ212760.1 |
- |
3, 4 |
277 |
|
FQ212769.1 |
- |
3, 4 |
277 |
|
FQ213060.1 |
- |
3, 4 |
273 |
|
FQ213396.1 |
- |
3, 4 |
273 |
|
FQ213533.1 |
- |
3, 4 |
280 |
|
FQ213898.1 |
- |
3, 4 |
269 |
|
FQ214072.1 |
- |
3, 4 |
277 |
|
FQ214282.1 |
- |
3, 4 |
277 |
|
FQ218386.1 |
- |
3, 4 |
276 |
|
FQ218473.1 |
- |
3, 4 |
306 |
|
FQ218485.1 |
- |
3, 4 |
277 |
|
FQ218540.1 |
- |
3, 4 |
277 |
|
FQ218699.1 |
- |
3, 4 |
279 |
|
FQ218898.1 |
- |
3, 4 |
279 |
|
FQ218953.1 |
- |
2, 3 |
333 |
|
FQ219009.1 |
- |
3, 4 |
277 |
|
FQ219124.1 |
- |
2, 3 |
237 |
|
FQ219144.1 |
- |
3, 4 |
277 |
|
FQ219202.1 |
- |
2, 3 |
228 |
|
FQ219319.1 |
- |
3, 4 |
277 |
|
FQ219432.1 |
- |
3, 4 |
277 |
|
FQ219448.1 |
- |
3, 4 |
279 |
|
FQ219475.1 |
- |
3, 4 |
272 |
|
FQ219490.1 |
- |
3, 4 |
277 |
|
FQ219516.1 |
- |
3, 4 |
277 |
|
FQ219534.1 |
- |
3, 4 |
277 |
|
FQ219700.1 |
- |
3, 4 |
277 |
|
FQ219711.1 |
- |
3, 4 |
277 |
|
FQ220125.1 |
- |
3, 4 |
273 |
|
FQ220532.1 |
- |
3, 4 |
273 |
|
FQ220683.1 |
- |
3, 4 |
277 |
|
FQ220752.1 |
- |
3, 4 |
308 |
|
FQ220879.1 |
- |
3, 4 |
281 |
|
FQ221856.1 |
- |
3, 4 |
277 |
|
FQ223299.1 |
- |
2, 3 |
228 |
|
FQ229186.1 |
- |
3, 4 |
277 |
|
FQ229201.1 |
- |
3, 4 |
277 |
|
FQ229530.1 |
- |
2, 3 |
228 |
|
FQ229565.1 |
- |
3, 4 |
273 |
|
FQ229650.1 |
- |
3, 4 |
277 |
|
FQ229759.1 |
- |
3, 4 |
277 |
|
FQ229950.1 |
- |
3, 4 |
277 |
|
FQ230084.1 |
- |
3, 4 |
277 |
|
FQ231375.1 |
- |
3, 4 |
306 |
|
J00705.1 |
- |
2, 3 |
235 |
|
View Full Product Details