Gene Transcripts

Gene Symbol
Nradd
Entrez Gene ID
246143
Gene Name
neurotrophin receptor associated death domain
Gene Aliases
NRH2, PLAIDD
Chromosome Location
Chr. 8: 118890402 - 118893712 on Build Rnor_6.0
UniGene ID
Rn.40242
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139259.2 NP_640352.1 3 265
GenBank mRNA AF497263.1 - 3 269
AF534395.1 - 2 81

View Full Product Details