Gene Transcripts

Gene Symbol
Retsat
Entrez Gene ID
246298
Gene Name
retinol saturase (all trans retinol 13,14 reductase)
Gene Aliases
Rmt7
Chromosome Location
Chr. 4: 100465365 - 100474128 on Build Rnor_6.0
UniGene ID
Rn.55275
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145084.1 NP_659552.1 1 152
GenBank mRNA AF465614.1 - 1 152

View Full Product Details