Gene Transcripts

Gene Symbol
Slc13a5
Entrez Gene ID
266998
Gene Name
solute carrier family 13 (sodium-dependent citrate transporter), member 5
Gene Aliases
Nact
Chromosome Location
Chr. 10: 58810054 - 58835475 on Build Rnor_6.0
UniGene ID
Rn.107996
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170668.1 NP_733768.1 2 141
XM_006246602.2 XP_006246664.1 3 1367
GenBank mRNA AF522186.1 - 2 141

View Full Product Details