Gene Transcripts
- Gene Symbol
- F7
- Entrez Gene ID
- 260320
- Gene Name
- coagulation factor VII (serum prothrombin conversion accelerator)
- Gene Aliases
- Chromosome Location
- Chr. 16: 81824610 - 81834923 on Build Rnor_6.0
- UniGene ID
- Rn.86416
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152846.1 |
NP_690059.1
|
6, 7 |
632 |
|
GenBank mRNA |
AF532184.1 |
- |
6, 7 |
632 |
|
View Full Product Details