Gene Transcripts
- Gene Symbol
- Ctsm
- Entrez Gene ID
- 306720
- Gene Name
- cathepsin M
- Gene Aliases
- Chromosome Location
- Chr. 17: 3820606 - 3826015 on Build Rnor_6.0
- UniGene ID
- Rn.23458
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181378.2 |
NP_852043.1
|
2, 3 |
200 |
|
GenBank mRNA |
AF456462.1 |
- |
2, 3 |
200 |
|
- Gene Symbol
- Cts8
- Entrez Gene ID
- 680718
- Gene Name
- cathepsin 8
- Gene Aliases
- Chromosome Location
- Chr. 17: 3385851 - 3397621 on Build Rnor_6.0
- UniGene ID
- Rn.220599
- Species
- Rattus norvegicus
- Gene Symbol
- RGD1564827
- Entrez Gene ID
- 498689
- Gene Name
- similar to cathepsin M
- Gene Aliases
- Chromosome Location
- Chr. 17: 4119288 - 4150720 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details