Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020062.3 | NP_064446.2 | 4 | 597 | |
GenBank mRNA | AB044777.1 | - | 3 | 488 | |
AB044786.1 | - | 2 | 158 | ||
AB052777.1 | - | 4 | 641 | ||
AF249267.3 | - | 4 | 542 | ||
AK298304.1 | - | 2 | 520 | ||
BC001402.2 | - | 3 | 368 | ||
BC052306.1 | - | 4 | 642 | 6576325 |