Gene Transcripts

Gene Symbol
Smpx
Entrez Gene ID
84416
Gene Name
small muscle protein, X-linked
Gene Aliases
Chromosome Location
Chr. X: 40030244 - 40086870 on Build Rnor_6.0
UniGene ID
Rn.4123
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053395.1 NP_445847.1 4 353
XM_006256945.2 XP_006257007.1 4 322
GenBank mRNA AF364071.1 - 4 353

View Full Product Details