Gene Transcripts

Gene Symbol
Ly6i
Entrez Gene ID
246138
Gene Name
lymphocyte antigen 6 complex, locus I
Gene Aliases
Ly6b
Chromosome Location
Chr. 7: 116419573 - 116425017 on Build Rnor_6.0
UniGene ID
Rn.39703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139257.1 NP_640350.1 3 267
XM_006241765.2 XP_006241827.1 254
XM_006241766.2 XP_006241828.1 4 394
XM_008765545.1 XP_008763767.1 4 381
GenBank mRNA M30689.1 - 3 267

View Full Product Details