Gene Transcripts

Gene Symbol
Ly6i
Entrez Gene ID
246138
Gene Name
lymphocyte antigen 6 complex, locus I
Gene Aliases
Ly6b
Chromosome Location
Chr. 7: 116419573 - 116425017 on Build Rnor_6.0
UniGene ID
Rn.39703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139257.1 NP_640350.1 3, 4 288
XM_006241765.2 XP_006241827.1 275
XM_006241766.2 XP_006241828.1 4, 5 415
XM_008765545.1 XP_008763767.1 4, 5 402
GenBank mRNA M30689.1 - 3, 4 288

Gene Symbol
Ly6al
Entrez Gene ID
362935
Gene Name
lymphocyte antigen 6 complex, locus A-like
Gene Aliases
Ly6a
Chromosome Location
Chr. 7: 116549110 - 116561263 on Build Rnor_6.0
UniGene ID
Rn.203167
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128099.1 NP_001121571.1 2, 3 186

View Full Product Details