Gene Transcripts

Gene Symbol
Nr2f2
Entrez Gene ID
113984
Gene Name
nuclear receptor subfamily 2, group F, member 2
Gene Aliases
Tfcoup2
Chromosome Location
Chr. 1: 131447671 - 131465745 on Build Rnor_6.0
UniGene ID
Rn.164334
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080778.2 NP_542956.1 1 492
GenBank mRNA AF003944.1 - 1 493

View Full Product Details