Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287389.1 | NP_001274318.1 | 7, 8 | 1135 | |
NM_007781.3 | NP_031807.1 | 9, 10 | 1462 | ||
GenBank mRNA | AK154650.1 | - | 9, 10 | 1385 | |
AK155216.1 | - | 9, 10 | 1388 | ||
AK155511.1 | - | 9, 10 | 1364 | ||
AK155626.1 | - | 7, 8 | 1037 | ||
AK155675.1 | - | 9, 10 | 1364 | ||
AK170028.1 | - | 9, 10 | 1364 | ||
M29855.1 | - | 9, 10 | 1344 |