Gene Transcripts

Gene Symbol
Cyp2b13
Entrez Gene ID
13089
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 13
Gene Aliases
Chromosome Location
Chr. 7: 26061344 - 26096197 on Build GRCm38
UniGene ID
Mm.482696
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007813.2 NP_031839.2 5 750
GenBank mRNA BC145895.1 - 5 748

View Full Product Details