Gene Transcripts

Gene Symbol
Hcls1
Entrez Gene ID
15163
Gene Name
hematopoietic cell specific Lyn substrate 1
Gene Aliases
AW213261, HS1
Chromosome Location
Chr. 16: 36934983 - 36963214 on Build GRCm38
UniGene ID
Mm.4091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008225.2 NP_032251.2 7 595
GenBank mRNA AK088723.1 - 7 577
BC007469.1 - 7 565 3593456
D42120.1 - 7 526
X84797.1 - 7 563

View Full Product Details