Gene Transcripts

Gene Symbol
Lta4h
Entrez Gene ID
16993
Gene Name
leukotriene A4 hydrolase
Gene Aliases
Chromosome Location
Chr. 10: 93453396 - 93484898 on Build GRCm38
UniGene ID
Mm.271071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008517.2 NP_032543.2 2 372
GenBank mRNA AK032828.1 - 2 317
AK134931.1 - 2 358
AK180384.1 - 2 142
BC021417.1 - 2 366 5004113
M63848.1 - 338

View Full Product Details