Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008798.2 | NP_032824.1 | 2 | 254 | |
GenBank mRNA | AK039828.1 | - | 2 | 234 | |
BC119179.1 | - | 2 | 223 | 8733928 | |
BC120602.1 | - | 2 | 223 | 40129525 | |
X67914.1 | - | 2 | 254 |