Gene Transcripts

Gene Symbol
Prkar2a
Entrez Gene ID
19087
Gene Name
protein kinase, cAMP dependent regulatory, type II alpha
Gene Aliases
1110061A24Rik, AI317181, AI836829, RII(alpha)
Chromosome Location
Chr. 9: 108692143 - 108749512 on Build GRCm38
UniGene ID
Mm.253102
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008924.2 NP_032950.1 1 460
GenBank mRNA AF533977.1 - 1 460
AK013274.1 - 1 450
AK132728.1 - 1 399
AK132824.1 - 1 396
BC075623.1 - 1 431 6854057
BC080276.1 - 1 454 30354403
J02935.1 - 1 143

View Full Product Details