Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008964.4 | NP_032990.1 | 1 | 1047 | |
GenBank mRNA | AK036474.1 | - | 1 | 492 | |
AK036531.1 | - | 1 | 966 | ||
AK054132.1 | - | 1 | 568 | ||
AK086904.1 | - | 1 | 961 | ||
AK136577.1 | - | 1 | 60 | ||
AK137515.1 | - | 1 | 257 | ||
BC005440.1 | - | 1 | 555 | 3487087 | |
D50589.1 | - | 1 | 338 |