Gene Transcripts

Gene Symbol
Siah1a
Entrez Gene ID
20437
Gene Name
seven in absentia 1A
Gene Aliases
AA982064, AI853500, Sinh1a
Chromosome Location
Chr. 8: 86723920 - 86746006 on Build GRCm38
UniGene ID
Mm.324553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009172.2 NP_033198.1 2 1601
GenBank mRNA AK019266.1 - 1 399
Z19579.1 - 2 1529

View Full Product Details