Gene Transcripts

Gene Symbol
CDC25B
Entrez Gene ID
994
Gene Name
cell division cycle 25B
Gene Aliases
Chromosome Location
Chr. 20: 3786772 - 3806121 on Build GRCh38
UniGene ID
Hs.153752
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287516.1 NP_001274445.1 8 974
NM_001287517.1 NP_001274446.1 8 932
NM_001287518.1 NP_001274447.1 8 974
NM_001287519.1 NP_001274448.1 8 705
NM_001287520.1 NP_001274449.1 8 747
NM_001287522.1 NP_001274451.1 7 624
NM_001287524.1 NP_001274453.1 4 529
NM_004358.4 NP_004349.1 8 1557
NM_021872.3 NP_068658.1 7 1476
NM_021873.3 NP_068659.1 8 1599
NR_136335.1 8 747
NR_136336.1 8 974
GenBank mRNA AK092713.1 - 8 747
AK295573.1 - 8 816
AK299028.1 - 8 781
AK299192.1 - 8 750
BC006395.2 - 8 880 4135465
BC009953.2 - 8 960 4130804
BC051711.1 - 8 1504 5301205
BX640836.1 - 4 521
BX647988.1 - 8 662
DA065196.1 - 4 529
FR695900.1 - 8 747
FR695901.1 - 7 624
M81934.1 - 8 836
S78187.1 - 8 1004
Z68092.1 - 7 784

View Full Product Details