Gene Transcripts

Gene Symbol
Ttk
Entrez Gene ID
22137
Gene Name
Ttk protein kinase
Gene Aliases
AL022661, Esk1, Mps1, PYT, esk
Chromosome Location
Chr. 9: 83834651 - 83872390 on Build GRCm38
UniGene ID
Mm.1904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110265.1 NP_001103735.1 14, 15 1630
NM_001284272.1 NP_001271201.1 14, 15 1630
NM_009445.2 NP_033471.2 14, 15 1630
GenBank mRNA AK041487.1 - 14, 15 1601
AK049200.1 - 14, 15 1625
AK164093.1 - 14, 15 1629
AX775731.1 - 14, 15 1596
BC058851.1 - 14, 15 1616 30023533
DV659530.1 - 1, 2 56
M86377.1 - 15, 16 1656

View Full Product Details