Gene Transcripts

Gene Symbol
Akr1b3
Entrez Gene ID
11677
Gene Name
aldo-keto reductase family 1, member B3 (aldose reductase)
Gene Aliases
ALR2, AR, Ahr-1, Ahr1, Akr1b1, Aldor1, Aldr1
Chromosome Location
Chr. 6: 34303930 - 34317489 on Build GRCm38
UniGene ID
Mm.389126
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009658.3 NP_033788.3 2 182
GenBank mRNA AK028079.1 - 2 137
AK138000.1 - 2 135
AK139250.1 - 2 137
AK145392.1 - 2 138
AK149861.1 - 2 172
AK150197.1 - 2 137
AK153949.1 - 2 166
AK164682.1 - 2 137
AK170663.1 - 2 184
AK192881.1 - 2 110
BC004725.1 - 2 129 3582788
BC021655.1 - 2 175 3708927
BC085310.1 - 2 114 6485416
BY749711.1 - 2 182
D32250.1 - 2 123
L39795.1 - 2 114
U29152.1 - 2 133

View Full Product Details