Gene Transcripts

Gene Symbol
Zfp92
Entrez Gene ID
22754
Gene Name
zinc finger protein 92
Gene Aliases
Chromosome Location
Chr. X: 73410978 - 73429813 on Build GRCm38
UniGene ID
Mm.5060
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009566.4 NP_033592.2 4 279
GenBank mRNA AK141522.1 - 4 280
BC096436.1 - 3 330 30478238
U47104.1 - 4 299

View Full Product Details