Gene Transcripts

Gene Symbol
Aoc3
Entrez Gene ID
11754
Gene Name
amine oxidase, copper containing 3
Gene Aliases
SSAO, VAP1
Chromosome Location
Chr. 11: 101330606 - 101339430 on Build GRCm38
UniGene ID
Mm.67281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009675.2 NP_033805.1 1 1385
GenBank mRNA AF054831.1 - 1 1400
AF115411.1 - 1 1216
BC080857.1 - 1 1224 6854465

View Full Product Details