Gene Transcripts
- Gene Symbol
- Cdc7
- Entrez Gene ID
- 12545
- Gene Name
- cell division cycle 7 (S. cerevisiae)
- Gene Aliases
- AI597260, Cdc7l1, muCdc7
- Chromosome Location
- Chr. 5: 106964315 - 106984440 on Build GRCm38
- UniGene ID
- Mm.20842
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271566.1 |
NP_001258495.1
|
3, 4 |
531 |
|
NM_001271567.1 |
NP_001258496.1
|
3, 4 |
362 |
|
NM_001271568.1 |
NP_001258497.1
|
3, 4 |
362 |
|
NM_009863.3 |
NP_033993.2
|
3, 4 |
362 |
|
GenBank mRNA |
AB018574.1 |
- |
3, 4 |
363 |
|
AB018575.1 |
- |
3, 4 |
363 |
|
AB018576.1 |
- |
2, 3 |
173 |
|
AB018577.1 |
- |
2, 3 |
173 |
|
AJ007661.1 |
- |
3, 4 |
314 |
|
AK012986.1 |
- |
3, 4 |
345 |
|
AK145921.1 |
- |
3, 4 |
288 |
|
AK146009.1 |
- |
3, 4 |
533 |
|
AK163921.1 |
- |
|
332 |
|
BC062149.1 |
- |
3, 4 |
292 |
|
BC072666.1 |
- |
3, 4 |
303 |
30611704 |
BC080702.1 |
- |
3, 4 |
315 |
30474846 |
View Full Product Details