Gene Transcripts

Gene Symbol
Ackr1
Entrez Gene ID
13349
Gene Name
atypical chemokine receptor 1 (Duffy blood group)
Gene Aliases
AA162249, CCBP1, CD234, Darc, Dfy, ESTM35, FY, GPD
Chromosome Location
Chr. 1: 173331886 - 173333503 on Build GRCm38
UniGene ID
Mm.6393
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF016584.1 - 2 836
AF109159.1 - 2 895
AY289754.1 - 2 802
AY291295.1 - 2 802
BC005583.1 - 2 894 3968984

View Full Product Details