Gene Transcripts
- Gene Symbol
- STYX
- Entrez Gene ID
- 6815
- Gene Name
- serine/threonine/tyrosine interacting protein
- Gene Aliases
- Chromosome Location
- Chr. 14: 52730159 - 52774989 on Build GRCh38
- UniGene ID
- Hs.364980
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130701.1 |
NP_001124173.1
|
3, 4 |
160 |
|
NM_145251.3 |
NP_660294.1
|
2, 3 |
408 |
|
XM_011537108.2 |
XP_011535410.1
|
2, 3 |
75 |
|
GenBank mRNA |
AK098195.1 |
- |
2, 3 |
391 |
|
AL832591.1 |
- |
3, 4 |
145 |
|
BC020265.2 |
- |
2, 3 |
359 |
3922829 |
BC146995.1 |
- |
2, 3 |
298 |
9056923 |
BC146998.1 |
- |
2, 3 |
298 |
9056926 |
BX647636.1 |
- |
2, 3 |
366 |
|
DB498442.1 |
- |
2, 3 |
411 |
|
View Full Product Details