Gene Transcripts
- Gene Symbol
- Hexa
- Entrez Gene ID
- 15211
- Gene Name
- hexosaminidase A
- Gene Aliases
- Hex-1
- Chromosome Location
- Chr. 9: 59539545 - 59565109 on Build GRCm38
- UniGene ID
- Mm.2284
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010421.4 |
NP_034551.2
|
4, 5 |
518 |
|
GenBank mRNA |
AK075895.1 |
- |
4, 5 |
494 |
|
AK075911.1 |
- |
4, 5 |
519 |
|
AK080777.1 |
- |
4, 5 |
491 |
|
AK144168.1 |
- |
4, 5 |
646 |
|
AK150444.1 |
- |
4, 5 |
489 |
|
AK151960.1 |
- |
4, 5 |
493 |
|
AK159091.1 |
- |
4, 5 |
510 |
|
AK159814.1 |
- |
4, 5 |
504 |
|
AK159890.1 |
- |
4, 5 |
491 |
|
AK160112.1 |
- |
4, 5 |
478 |
|
AK167013.1 |
- |
4, 5 |
504 |
|
AK168186.1 |
- |
4, 5 |
518 |
|
AK170903.1 |
- |
4, 5 |
504 |
|
BC010755.1 |
- |
4, 5 |
487 |
4225233 |
U07631.1 |
- |
4, 5 |
510 |
|
X64331.1 |
- |
4, 5 |
461 |
|
View Full Product Details