Gene Transcripts
- Gene Symbol
- Ica1
- Entrez Gene ID
- 15893
- Gene Name
- islet cell autoantigen 1
- Gene Aliases
- 69kDa, ICA69
- Chromosome Location
- Chr. 6: 8630527 - 8778537 on Build GRCm38
- UniGene ID
- Mm.275683
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252266.1 |
NP_001239195.1
|
8, 9 |
977 |
|
NM_010492.3 |
NP_034622.3
|
9, 10 |
1390 |
|
GenBank mRNA |
AB303567.1 |
- |
8, 9 |
901 |
|
BC002030.1 |
- |
9, 10 |
1024 |
3487814 |
CB056959.1 |
- |
4, 5 |
487 |
|
DV658174.1 |
- |
3, 4 |
300 |
|
DV659030.1 |
- |
7, 8 |
763 |
|
U26459.1 |
- |
9, 10 |
1013 |
|
U26460.1 |
- |
8, 9 |
919 |
|
U26461.1 |
- |
8, 9 |
918 |
|
U34595.1 |
- |
7, 8 |
886 |
|
View Full Product Details