Gene Transcripts

Gene Symbol
Insr
Entrez Gene ID
16337
Gene Name
insulin receptor
Gene Aliases
4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
Chromosome Location
Chr. 8: 3150922 - 3279649 on Build GRCm38
UniGene ID
Mm.268003
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010568.2 NP_034698.2 2 634
GenBank mRNA AK052187.1 - 2 569
J05149.1 - 2 193
KC356871.1 - 2 145

View Full Product Details