Gene Transcripts

Gene Symbol
Itpr1
Entrez Gene ID
16438
Gene Name
inositol 1,4,5-trisphosphate receptor 1
Gene Aliases
D6Pas2, ENSMUSG00000072853, Gm10429, IP3R1, InsP3R, Ip3r, Itpr-1, P400, Pcp-1, Pcp1, opt
Chromosome Location
Chr. 6: 108213083 - 108551116 on Build GRCm38
UniGene ID
Mm.227912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010585.5 NP_034715.3 7 830
GenBank mRNA AK047719.1 - 7 831
AK049015.1 - 4 444
AK137477.1 - 7 832
X15373.1 - 7 808

View Full Product Details